Learn More
Meso Scale Discovery V-PLEX NHP IL-8 Kit
SDP
Catalog No. 5000080831
Encompass
The V-PLEX NHP IL-8 Kit from Meso Scale Discovery is an electrochemiluminescent immunoassay for the quantitative measurement of human interleukin 8 (IL-8) in cell culture supernatants serum plasma and other biological samples The kit contains a 96-well plate coated with a monoclonal antibody to human IL-8 a detection antibody conjugated to an electrochemiluminescent label calibrators diluent wash buffer read buffer and plate seals The assay has a sensitivity of 0 09 pg/mL and a 7-log dynamic range from 0 09-10 000 pg/mL The total assay time is 3 5 hours This kit allows for the accurate quantitation of IL-8 levels in human samples
Catalog No. 50-000-80831 Supplier Meso Scale Discovery Supplier No. K156RAD4
Add to Cart
Add to Cart
Request a Quote
The V-PLEX NHP IL-8 Kit from Meso Scale Discovery is an electrochemiluminescent immunoassay for the quantitative measurement of human interleukin 8 (IL-8) in cell culture supernatants serum plasma and other biological samples The kit contains a 96-well plate coated with a monoclonal antibody to human IL-8 a detection antibody conjugated to an electrochemiluminescent label calibrators diluent wash buffer read buffer and plate seals The assay has a sensitivity of 0 09 pg/mL and a 7-log dynamic range from 0 09-10 000 pg/mL The total assay time is 3 5 hours This kit allows for the accurate quantitation of IL-8 levels in human samples