Learn More
Meso Scale Discovery V-PLEX NHP IL-8 Kit
SDP
Catalog No. 5000081141
Encompass
The V-PLEX NHP IL-8 Kit from Meso Scale Discovery is a multiplex immunoassay kit that allows for the quantitative measurement of human IL-8 in serum plasma and cell culture supernatants The kit utilizes electrochemiluminescent detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent labels calibrators diluent wash buffer read buffer and plate seals The assay has a typical sensitivity of 0 04 pg/mL and an upper limit of quantification of 2500 pg/mL
Catalog No. 50-000-81141 Supplier Meso Scale Discovery Supplier No. K156RAD1
The V-PLEX NHP IL-8 Kit from Meso Scale Discovery is a multiplex immunoassay kit that allows for the quantitative measurement of human IL-8 in serum plasma and cell culture supernatants The kit utilizes electrochemiluminescent detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent labels calibrators diluent wash buffer read buffer and plate seals The assay has a typical sensitivity of 0 04 pg/mL and an upper limit of quantification of 2500 pg/mL