Learn More
Meso Scale Discovery V-PLEX NHP IL-8 Kit
SDP
Catalog No. 5000081146
Encompass
The Meso Scale Discovery V-PLEX NHP IL-8 Kit is a sensitive immunoassay for measuring human interleukin 8 (IL-8) in serum plasma and culture supernatant It uses MSD s SULFO-TAG labels and multi-array technology for superior sensitivity wide dynamic range and quick results The kit includes a 96-well plate with capture antibodies detection antibodies with ruthenium labels calibrators and necessary reagents It detects as low as 0 04 pg/mL and up to 10 000 pg/mL ideal for various research applications
Catalog No. 50-000-81146 Supplier Meso Scale Discovery Supplier No. K156RAD2
The Meso Scale Discovery V-PLEX NHP IL-8 Kit is a sensitive immunoassay for measuring human interleukin 8 (IL-8) in serum plasma and culture supernatant It uses MSD s SULFO-TAG labels and multi-array technology for superior sensitivity wide dynamic range and quick results The kit includes a 96-well plate with capture antibodies detection antibodies with ruthenium labels calibrators and necessary reagents It detects as low as 0 04 pg/mL and up to 10 000 pg/mL ideal for various research applications