Learn More
Meso Scale Discovery V-PLEX NHP MCP-1 Kit
SDP
Catalog No. 5000079019
Encompass
The V-PLEX NHP MCP-1 Kit from Meso Scale Discovery is a multiplex immunoassay kit that allows for the simultaneous quantitative measurement of human Monocyte Chemoattractant Protein 1 (MCP-1) in serum plasma and cell culture supernatants The kit utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity a broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with an ECL compound calibrators diluents wash buffers substrates and plate seals The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantitation of 10 000 pg/mL
Catalog No. 50-000-79019 Supplier Meso Scale Discovery Supplier No. K156NND2
Add to Cart
Add to Cart
The V-PLEX NHP MCP-1 Kit from Meso Scale Discovery is a multiplex immunoassay kit that allows for the simultaneous quantitative measurement of human Monocyte Chemoattractant Protein 1 (MCP-1) in serum plasma and cell culture supernatants The kit utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity a broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with an ECL compound calibrators diluents wash buffers substrates and plate seals The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantitation of 10 000 pg/mL