Learn More
Meso Scale Discovery V-PLEX NHP MCP-1 Kit
SDP
Catalog No. 5000079920
Encompass
The V-PLEX NHP MCP-1 Kit from Meso Scale Discovery is a multiplex immunoassay kit that enables the simultaneous quantitative measurement of human Monocyte Chemoattractant Protein 1 (MCP-1) in serum plasma and cell culture supernatants The kit utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This assay requires 50 L or less sample volume and has a testing time of 3 5 hours The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with ruthenium read buffer wash buffer calibrators and controls
Catalog No. 50-000-79920 Supplier Meso Scale Discovery Supplier No. K156NND1
The V-PLEX NHP MCP-1 Kit from Meso Scale Discovery is a multiplex immunoassay kit that enables the simultaneous quantitative measurement of human Monocyte Chemoattractant Protein 1 (MCP-1) in serum plasma and cell culture supernatants The kit utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This assay requires 50 L or less sample volume and has a testing time of 3 5 hours The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with ruthenium read buffer wash buffer calibrators and controls