Learn More
Meso Scale Discovery V-PLEX NHP MCP-4 Kit
SDP
Catalog No. 5000081144
Encompass
The V-PLEX NHP MCP-4 Kit from Meso Scale Diagnostics is a multiplex immunoassay kit for the simultaneous quantitative measurement of 4 neurodegenerative biomarkers in human serum plasma and CSF samples The kit contains a 96-well plate coated with capture antibodies for the biomarkers A 42 t-tau p-tau and -synuclein It utilizes electrochemiluminescent detection for high sensitivity and a broad dynamic range The kit includes all necessary reagents and calibrators for performing the 4-plex assay It is intended for research use only The kit allows efficient profiling of key biomarkers associated with Alzheimer s and Parkinson s diseases
Catalog No. 50-000-81144 Supplier Meso Scale Discovery Supplier No. K156NOD1
Add to Cart
Add to Cart
The V-PLEX NHP MCP-4 Kit from Meso Scale Diagnostics is a multiplex immunoassay kit for the simultaneous quantitative measurement of 4 neurodegenerative biomarkers in human serum plasma and CSF samples The kit contains a 96-well plate coated with capture antibodies for the biomarkers A 42 t-tau p-tau and -synuclein It utilizes electrochemiluminescent detection for high sensitivity and a broad dynamic range The kit includes all necessary reagents and calibrators for performing the 4-plex assay It is intended for research use only The kit allows efficient profiling of key biomarkers associated with Alzheimer s and Parkinson s diseases