Learn More
Meso Scale Discovery V-PLEX NHP MCP-4 Kit
SDP
Catalog No. 5000081251
Encompass
The Meso Scale Discovery V-PLEX Human MCP-4 Kit is an electrochemiluminescent immunoassay for measuring monocyte chemoattractant protein 4 (MCP-4) in human serum plasma and cell culture supernatants It uses MSD s V-PLEX technology offering high sensitivity a broad dynamic range and minimal cross-reactivity The kit includes a 96-well plate with capture antibodies detection antibodies and calibrators Results are read on MSD s SECTOR Imager and analyzed with MSD Discovery Workbench software providing accurate MCP-4 quantitation in under 5 hours
Catalog No. 50-000-81251 Supplier Meso Scale Discovery Supplier No. K156NOD4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX Human MCP-4 Kit is an electrochemiluminescent immunoassay for measuring monocyte chemoattractant protein 4 (MCP-4) in human serum plasma and cell culture supernatants It uses MSD s V-PLEX technology offering high sensitivity a broad dynamic range and minimal cross-reactivity The kit includes a 96-well plate with capture antibodies detection antibodies and calibrators Results are read on MSD s SECTOR Imager and analyzed with MSD Discovery Workbench software providing accurate MCP-4 quantitation in under 5 hours