Learn More
Meso Scale Discovery V-PLEX NHP MDC Kit
SDP
Catalog No. 5000080771
Encompass
The V-PLEX NHP MDC Kit from Meso Scale Diagnostics is a multiplex assay kit that allows for the simultaneous quantitative measurement of human cytokines chemokines growth factors and immune modulators in serum plasma tissue culture supernatant and other biological fluids The kit contains a 96-well plate coated with capture antibodies for 10 analytes detection antibodies standards buffers substrate and plate seals The assay uses electrochemiluminescence for high sensitivity and broad dynamic range This kit allows for efficient and accurate profiling of natural and adaptive human immune responses
Catalog No. 50-000-80771 Supplier Meso Scale Discovery Supplier No. K156NPD1
The V-PLEX NHP MDC Kit from Meso Scale Diagnostics is a multiplex assay kit that allows for the simultaneous quantitative measurement of human cytokines chemokines growth factors and immune modulators in serum plasma tissue culture supernatant and other biological fluids The kit contains a 96-well plate coated with capture antibodies for 10 analytes detection antibodies standards buffers substrate and plate seals The assay uses electrochemiluminescence for high sensitivity and broad dynamic range This kit allows for efficient and accurate profiling of natural and adaptive human immune responses