Learn More
Meso Scale Discovery V-PLEX NHP MDC Kit
SDP
Catalog No. 5000080772
Encompass
The V-PLEX NHP MDC Kit from Meso Scale Diagnostics is a multiplex immunoassay for measuring human cytokines chemokines growth factors and immune modulators in serum plasma and culture supernatants It includes a 96-well plate with capture antibodies detection antibodies calibrators diluent read buffer wash buffer and plate seals It measures up to 10 analytes like IL-2 IL-4 IL-6 IL-10 IL-12p70 IL-13 IFN-gamma TNF-alpha MCP-1 and GM-CSF The electrochemiluminescent detection offers high sensitivity wide range and low sample volume enabling efficient analysis of multiple immune mediators
Catalog No. 50-000-80772 Supplier Meso Scale Discovery Supplier No. K156NPD4
Add to Cart
Add to Cart
Request a Quote
The V-PLEX NHP MDC Kit from Meso Scale Diagnostics is a multiplex immunoassay for measuring human cytokines chemokines growth factors and immune modulators in serum plasma and culture supernatants It includes a 96-well plate with capture antibodies detection antibodies calibrators diluent read buffer wash buffer and plate seals It measures up to 10 analytes like IL-2 IL-4 IL-6 IL-10 IL-12p70 IL-13 IFN-gamma TNF-alpha MCP-1 and GM-CSF The electrochemiluminescent detection offers high sensitivity wide range and low sample volume enabling efficient analysis of multiple immune mediators