Learn More
Meso Scale Discovery V-PLEX NHP MDC Kit
SDP
Catalog No. 5000081522
Encompass
The Meso Scale Discovery V-PLEX NHP MDC Kit is a multiplex immunoassay that simultaneously measures natural killer cell and dendritic cell biomarkers in human serum plasma and culture supernatants This kit includes a 96-well plate pre-coated with capture antibodies for the following analytes CD16 CD56 CD69 HLA-DR CD11c and CD123 The electrochemiluminescent detection method provides high sensitivity and a broad dynamic range This product requires a Meso Scale Discovery SECTOR Imager for analysis With a small sample volume requirement and rapid turnaround time this kit enables efficient profiling of natural killer and dendritic cell activity
Catalog No. 50-000-81522 Supplier Meso Scale Discovery Supplier No. K156NPD2
The Meso Scale Discovery V-PLEX NHP MDC Kit is a multiplex immunoassay that simultaneously measures natural killer cell and dendritic cell biomarkers in human serum plasma and culture supernatants This kit includes a 96-well plate pre-coated with capture antibodies for the following analytes CD16 CD56 CD69 HLA-DR CD11c and CD123 The electrochemiluminescent detection method provides high sensitivity and a broad dynamic range This product requires a Meso Scale Discovery SECTOR Imager for analysis With a small sample volume requirement and rapid turnaround time this kit enables efficient profiling of natural killer and dendritic cell activity