Learn More
Meso Scale Discovery V-PLEX NHP MIP-1a Kit
SDP
Catalog No. 5000079023
Encompass
The V-PLEX NHP MIP-1 Kit from Meso Scale Discovery is a multiplex immunoassay kit for measuring human Macrophage Inflammatory Protein 1 alpha (MIP-1 ) in various biological fluids It uses electrochemiluminescence (ECL) detection for high sensitivity and broad dynamic range with low sample volume The kit includes a 96-well plate with capture antibodies ruthenium-labeled detection antibodies buffers blocker solutions calibrators and controls Sensitivity is 0 3 pg/mL with an upper quantification limit of 10 000 pg/mL
Catalog No. 50-000-79023 Supplier Meso Scale Discovery Supplier No. K156NQD2
The V-PLEX NHP MIP-1 Kit from Meso Scale Discovery is a multiplex immunoassay kit for measuring human Macrophage Inflammatory Protein 1 alpha (MIP-1 ) in various biological fluids It uses electrochemiluminescence (ECL) detection for high sensitivity and broad dynamic range with low sample volume The kit includes a 96-well plate with capture antibodies ruthenium-labeled detection antibodies buffers blocker solutions calibrators and controls Sensitivity is 0 3 pg/mL with an upper quantification limit of 10 000 pg/mL