Learn More
Meso Scale Discovery V-PLEX NHP MIP-1a Kit
SDP
Catalog No. 5000079946
Encompass
The V-PLEX NHP MIP-1 Kit from Meso Scale Discovery is a multiplex immunoassay kit that enables the simultaneous quantitative measurement of Macrophage Inflammatory Protein 1-alpha (MIP-1 ) in human serum plasma and cell culture supernatants The kit utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent labels calibrators and diluents The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 10 000 pg/mL
Catalog No. 50-000-79946 Supplier Meso Scale Discovery Supplier No. K156NQD4
Add to Cart
Add to Cart
Request a Quote
The V-PLEX NHP MIP-1 Kit from Meso Scale Discovery is a multiplex immunoassay kit that enables the simultaneous quantitative measurement of Macrophage Inflammatory Protein 1-alpha (MIP-1 ) in human serum plasma and cell culture supernatants The kit utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent labels calibrators and diluents The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 10 000 pg/mL