Learn More
Meso Scale Discovery V-PLEX NHP MIP-1a Kit
SDP
Catalog No. 5000081523
Encompass
The V-PLEX NHP MIP-1 Kit from Meso Scale Discovery is a multiplex immunoassay kit for measuring human Macrophage Inflammatory Protein 1 alpha (MIP-1 ) in various biological fluids It includes a 96-well plate with capture antibodies detection antibodies with an electrochemiluminescent label calibrators diluent wash buffer read buffer and plate seals The assay detects 3 2-10 000 pg/mL with minimal cross-reactivity and provides high-sensitivity results from a small sample in 3 5 hours
Catalog No. 50-000-81523 Supplier Meso Scale Discovery Supplier No. K156NQD1
The V-PLEX NHP MIP-1 Kit from Meso Scale Discovery is a multiplex immunoassay kit for measuring human Macrophage Inflammatory Protein 1 alpha (MIP-1 ) in various biological fluids It includes a 96-well plate with capture antibodies detection antibodies with an electrochemiluminescent label calibrators diluent wash buffer read buffer and plate seals The assay detects 3 2-10 000 pg/mL with minimal cross-reactivity and provides high-sensitivity results from a small sample in 3 5 hours