Learn More
Meso Scale Discovery V-PLEX NHP TARC Kit
SDP
Catalog No. 5000080000
Encompass
The V-PLEX NHP TARC Kit from Meso Scale Diagnostics is a highly sensitive immunoassay that quantitatively measures TARC (thymus and activation regulated chemokine) in human serum plasma and cell culture supernatants This kit utilizes electrochemiluminescence detection technology and provides reagents for 96 tests in a quick 4 5 hour protocol The lower limit of detection is 1 5 pg/mL with an upper limit of 10 000 pg/mL This assay kit offers high sensitivity wide dynamic range and requires low sample volume It is useful for studying Th2-mediated inflammatory responses
Catalog No. 50-000-80000 Supplier Meso Scale Discovery Supplier No. K156NTD1
Add to Cart
Add to Cart
The V-PLEX NHP TARC Kit from Meso Scale Diagnostics is a highly sensitive immunoassay that quantitatively measures TARC (thymus and activation regulated chemokine) in human serum plasma and cell culture supernatants This kit utilizes electrochemiluminescence detection technology and provides reagents for 96 tests in a quick 4 5 hour protocol The lower limit of detection is 1 5 pg/mL with an upper limit of 10 000 pg/mL This assay kit offers high sensitivity wide dynamic range and requires low sample volume It is useful for studying Th2-mediated inflammatory responses