Learn More
Meso Scale Discovery V-PLEX NHP TARC Kit
SDP
Catalog No. 5000080775
Encompass
The Meso Scale Discovery V-PLEX NHP TARC Kit is a sensitive immunoassay for measuring Th2-associated chemokine receptor (TARC/CCL17) in human serum plasma and culture supernatants It uses MSD s SULFO-TAG labels and multi-array technology for accurate reproducible results with high sensitivity and a broad dynamic range The kit includes a 96-well plate with capture antibody detection antibody read buffer wash buffer blocker solutions calibrator and controls for TARC analysis aiding in understanding Th2-mediated conditions
Catalog No. 50-000-80775 Supplier Meso Scale Discovery Supplier No. K156NTD4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX NHP TARC Kit is a sensitive immunoassay for measuring Th2-associated chemokine receptor (TARC/CCL17) in human serum plasma and culture supernatants It uses MSD s SULFO-TAG labels and multi-array technology for accurate reproducible results with high sensitivity and a broad dynamic range The kit includes a 96-well plate with capture antibody detection antibody read buffer wash buffer blocker solutions calibrator and controls for TARC analysis aiding in understanding Th2-mediated conditions