Learn More
Meso Scale Discovery V-PLEX NHP TNF-SY Kit
SDP
Catalog No. 5000080810
Encompass
The V-PLEX NHP TNF-beta Kit from Meso Scale Discovery is an immunoassay that uses electrochemiluminescence (ECL) technology for the quantitative measurement of natural human TNF-beta in serum plasma and cell culture supernatants The assay has high sensitivity a broad dynamic range and requires a low sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with an ECL compound calibrators diluent wash buffer read buffer and plate seals The assay takes approximately 3 5 hours to complete
Catalog No. 50-000-80810 Supplier Meso Scale Discovery Supplier No. K156RGD2
The V-PLEX NHP TNF-beta Kit from Meso Scale Discovery is an immunoassay that uses electrochemiluminescence (ECL) technology for the quantitative measurement of natural human TNF-beta in serum plasma and cell culture supernatants The assay has high sensitivity a broad dynamic range and requires a low sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with an ECL compound calibrators diluent wash buffer read buffer and plate seals The assay takes approximately 3 5 hours to complete