Learn More
Meso Scale Discovery V-PLEX NHP VEGF Kit
SDP
Catalog No. 5000079914
Encompass
The V-PLEX NHP VEGF Kit from Meso Scale Discovery is a multiplex immunoassay kit that allows for the simultaneous quantitative measurement of VEGF-A VEGF-C and VEGF-D in human serum plasma and cell culture supernatants The kit utilizes electrochemiluminescent detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent labels calibrators diluent wash buffer read buffer and plate seals The assay has a typical runtime of 3 5 hours and shows minimal cross-reactivity between targets
Catalog No. 50-000-79914 Supplier Meso Scale Discovery Supplier No. K156RHD1
Add to Cart
Add to Cart
The V-PLEX NHP VEGF Kit from Meso Scale Discovery is a multiplex immunoassay kit that allows for the simultaneous quantitative measurement of VEGF-A VEGF-C and VEGF-D in human serum plasma and cell culture supernatants The kit utilizes electrochemiluminescent detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent labels calibrators diluent wash buffer read buffer and plate seals The assay has a typical runtime of 3 5 hours and shows minimal cross-reactivity between targets