Learn More
Meso Scale Discovery V-PLEX NHP VEGF Kit, 5 plates, 96-well format, Assay Type: V-PLEX, Analyte: VEGF-A, Sensitivity: 1.12 pg/mL, Dynamic Range: 1.12 - 562 pg/mL
SDP
Catalog No. NC3591285
Encompass
V-PLEX NHP VEGF Kit, 5 plates, 96-well format, Assay Type: V-PLEX, Analyte: VEGF-A, Sensitivity: 1.12 pg/mL, Dynamic Range: 1.12 - 562 pg/mL
Catalog No. NC3591285 Supplier Meso Scale Discovery Supplier No. K156RHD2
Add to Cart
Add to Cart
V-PLEX NHP VEGF Kit, 5 plates, 96-well format, Assay Type: V-PLEX, Analyte: VEGF-A, Sensitivity: 1.12 pg/mL, Dynamic Range: 1.12 - 562 pg/mL