Learn More
Meso Scale Discovery V-PLEX PLS HUM BMRKR 39PLX KIT, 39 analytes, multiplex immunoassay, electrochemiluminescence (ECL), wide linear dynamic range, lot-to-lot consistent, serum/plasma, 96-well plate
SDP
Catalog No. NC3661641
Encompass
The V-PLEX Plus Human Biomarker 39-Plex Kit is a validated multiplex immunoassay for the accurate measurement of 39 human biomarkers using MULTI-ARRAY technology and electrochemiluminescence (ECL) detection. Designed for rigorous applications, it offers exceptional sensitivity and reproducibility, suitable for inflammation, chemotaxis, angiogenesis, and immune system regulation studies.
  • Multiplex immunoassay for measuring multiple biomarkers
  • Highly sensitive detection limits in sub- to low pg/mL range
  • Wide linear dynamic range for quantifying diverse analyte levels
  • Analytically validated for consistent results across lots and runs
  • Saves time by analyzing multiple analytes from a small-volume sample
  • Includes essential components for ease of use
  • Universal instrument compatibility across leading platforms
  • Sample types validated: Serum, Plasma
  • Research use only
Catalog No. NC3661641 Supplier Meso Scale Discovery Supplier No. K15209G1
Add to Cart
Add to Cart
Request a Quote
The V-PLEX Plus Human Biomarker 39-Plex Kit is a validated multiplex immunoassay for the accurate measurement of 39 human biomarkers using MULTI-ARRAY technology and electrochemiluminescence (ECL) detection. Designed for rigorous applications, it offers exceptional sensitivity and reproducibility, suitable for inflammation, chemotaxis, angiogenesis, and immune system regulation studies.
  • Multiplex immunoassay for measuring multiple biomarkers
  • Highly sensitive detection limits in sub- to low pg/mL range
  • Wide linear dynamic range for quantifying diverse analyte levels
  • Analytically validated for consistent results across lots and runs
  • Saves time by analyzing multiple analytes from a small-volume sample
  • Includes essential components for ease of use
  • Universal instrument compatibility across leading platforms
  • Sample types validated: Serum, Plasma
  • Research use only