Learn More
Meso Scale Discovery V-PLEX Plus GIP active Kit
SDP
Catalog No. 5000081632
Encompass
The V-PLEX Plus GIP Active Kit from Meso Scale Diagnostics is a 96-well plate assay kit for the quantitative measurement of GIP in human serum plasma and cell culture supernatants The kit uses electrochemiluminescence (ECL) technology and includes a V-PLEX Human GIP Plate V-PLEX Detection Antibody Read Buffer and ancillary reagents The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 1000 pg/mL The total assay time is approximately 3 5 hours This product requires the MSD QuickPlex SQ 120 or MESO QuickPlex SQ 120 Instrument
Catalog No. 50-000-81632 Supplier Meso Scale Discovery Supplier No. K150T5G1
The V-PLEX Plus GIP Active Kit from Meso Scale Diagnostics is a 96-well plate assay kit for the quantitative measurement of GIP in human serum plasma and cell culture supernatants The kit uses electrochemiluminescence (ECL) technology and includes a V-PLEX Human GIP Plate V-PLEX Detection Antibody Read Buffer and ancillary reagents The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 1000 pg/mL The total assay time is approximately 3 5 hours This product requires the MSD QuickPlex SQ 120 or MESO QuickPlex SQ 120 Instrument