Learn More
Meso Scale Discovery V-PLEX Plus GLP-1 Active Kit
SDP
Catalog No. 5000081569
Encompass
The Meso Scale Discovery V-PLEX Plus GLP-1 Active Kit quantitatively measures active GLP-1 in serum plasma and cell culture supernatants This electrochemiluminescent immunoassay uses MSD s SULFO-TAG labels and multi-array technology for sensitive detection It includes a 96-well plate with a GLP-1 capture antibody a detection antibody with an MSD SULFO-TAG and calibrators and controls for a standard curve With a 2-hour sample incubation and minimal hands-on time it enables high-throughput quantification of active GLP-1
Catalog No. 50-000-81569 Supplier Meso Scale Discovery Supplier No. K1503OG2
The Meso Scale Discovery V-PLEX Plus GLP-1 Active Kit quantitatively measures active GLP-1 in serum plasma and cell culture supernatants This electrochemiluminescent immunoassay uses MSD s SULFO-TAG labels and multi-array technology for sensitive detection It includes a 96-well plate with a GLP-1 capture antibody a detection antibody with an MSD SULFO-TAG and calibrators and controls for a standard curve With a 2-hour sample incubation and minimal hands-on time it enables high-throughput quantification of active GLP-1