Learn More
Meso Scale Discovery V-PLEX Plus Glucagon Kit
SDP
Catalog No. 5000081782
Encompass
The Meso Scale Discovery V-PLEX Plus Glucagon Kit is a sensitive immunoassay for measuring glucagon in human serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array plate technology for rapid accurate glucagon measurement with high sensitivity and a broad dynamic range The kit includes a 96-well plate with capture antibody detection antibody with ruthenium label calibrators and necessary reagents With a detection limit of 0 24 pg/mL it s ideal for studying glucagon secretion and its role in glucose homeostasis
Catalog No. 50-000-81782 Supplier Meso Scale Discovery Supplier No. K150U5G2
The Meso Scale Discovery V-PLEX Plus Glucagon Kit is a sensitive immunoassay for measuring glucagon in human serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array plate technology for rapid accurate glucagon measurement with high sensitivity and a broad dynamic range The kit includes a 96-well plate with capture antibody detection antibody with ruthenium label calibrators and necessary reagents With a detection limit of 0 24 pg/mL it s ideal for studying glucagon secretion and its role in glucose homeostasis