Learn More
Meso Scale Discovery V-PLEX Plus Human Ab42 Kit
SDP
Catalog No. 5000079160
Encompass
The Meso Scale Discovery V-PLEX Plus Human A 42 Kit is an electrochemiluminescent immunoassay for measuring A 42 in human CSF serum and plasma It uses MSD s SULFO-TAG labels and multi-array technology for a sensitive assay detecting A 42 as low as 1 6 pg/mL The kit includes a 96-well plate with capture antibody detection antibody with SULFO-TAG calibrators and diluent The assay range is 1 6-1000 pg/mL and takes about 3 5 hours This kit quantifies A 42 a key Alzheimer s biomarker
Catalog No. 50-000-79160 Supplier Meso Scale Discovery Supplier No. K151LBG4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX Plus Human A 42 Kit is an electrochemiluminescent immunoassay for measuring A 42 in human CSF serum and plasma It uses MSD s SULFO-TAG labels and multi-array technology for a sensitive assay detecting A 42 as low as 1 6 pg/mL The kit includes a 96-well plate with capture antibody detection antibody with SULFO-TAG calibrators and diluent The assay range is 1 6-1000 pg/mL and takes about 3 5 hours This kit quantifies A 42 a key Alzheimer s biomarker