Learn More
Meso Scale Discovery V-PLEX Plus Human Ab42 Kit
SDP
Catalog No. 5000081177
Encompass
The V-PLEX Plus Human A 42 Kit from Meso Scale Discovery is an immunoassay that uses electrochemiluminescence (ECL) for the quantitative measurement of amyloid beta 1-42 (A 42) in human cerebrospinal fluid (CSF) serum and plasma It utilizes a V-PLEX sandwich immunoassay format with a monoclonal antibody specific for the N-terminus of A 42 coated on carbon electrodes in 96-well plates Samples and calibrators are added followed by a detection antibody conjugated with an ECL label Levels of A 42 are quantified using an MSD instrument by measuring ECL signals The kit includes a plate detection antibody calibrator diluent and wash buffer for 96 tests with a testing time of 3 5 hours
Catalog No. 50-000-81177 Supplier Meso Scale Discovery Supplier No. K151LBG2
Add to Cart
Add to Cart
Request a Quote
The V-PLEX Plus Human A 42 Kit from Meso Scale Discovery is an immunoassay that uses electrochemiluminescence (ECL) for the quantitative measurement of amyloid beta 1-42 (A 42) in human cerebrospinal fluid (CSF) serum and plasma It utilizes a V-PLEX sandwich immunoassay format with a monoclonal antibody specific for the N-terminus of A 42 coated on carbon electrodes in 96-well plates Samples and calibrators are added followed by a detection antibody conjugated with an ECL label Levels of A 42 are quantified using an MSD instrument by measuring ECL signals The kit includes a plate detection antibody calibrator diluent and wash buffer for 96 tests with a testing time of 3 5 hours