Learn More
Meso Scale Discovery V-PLEX Plus Human CRP Kit
SDP
Catalog No. 5000078516
Encompass
The V-PLEX Plus Human CRP Kit from Meso Scale Diagnostics is an electrochemiluminescent immunoassay for the quantitative measurement of C-reactive protein (CRP) in human serum plasma and cell culture supernatants The kit contains a 96-well plate coated with capture antibody detection antibody conjugated to an electrochemiluminescent label calibrators diluent wash buffer read buffer and plate seals The assay has a range of 0 016-50 ng/mL and takes approximately 3 5 hours to complete It utilizes MSD s proprietary SECTOR Imager for high sensitivity and a broad dynamic range The kit provides a simple rapid and sensitive method to quantify CRP levels
Catalog No. 50-000-78516 Supplier Meso Scale Discovery Supplier No. K151STG1
The V-PLEX Plus Human CRP Kit from Meso Scale Diagnostics is an electrochemiluminescent immunoassay for the quantitative measurement of C-reactive protein (CRP) in human serum plasma and cell culture supernatants The kit contains a 96-well plate coated with capture antibody detection antibody conjugated to an electrochemiluminescent label calibrators diluent wash buffer read buffer and plate seals The assay has a range of 0 016-50 ng/mL and takes approximately 3 5 hours to complete It utilizes MSD s proprietary SECTOR Imager for high sensitivity and a broad dynamic range The kit provides a simple rapid and sensitive method to quantify CRP levels