Learn More
Meso Scale Discovery V-PLEX Plus Human CRP Kit
SDP
Catalog No. 5000079794
Encompass
The V-PLEX Plus Human CRP Kit from Meso Scale Discovery is an electrochemiluminescent immunoassay for measuring C-reactive protein (CRP) in human serum plasma and cell culture supernatants It includes a 96-well plate with capture antibody detection antibody with an electrochemiluminescent label calibrators diluent wash buffer read buffer and plate seals The assay range is 0 016-50 ng/mL with 0 008 ng/mL sensitivity requiring only 50 L of sample It processes up to 94 samples in duplicate with high sensitivity and a wide dynamic range
Catalog No. 50-000-79794 Supplier Meso Scale Discovery Supplier No. K151STG4
Add to Cart
Add to Cart
Request a Quote
The V-PLEX Plus Human CRP Kit from Meso Scale Discovery is an electrochemiluminescent immunoassay for measuring C-reactive protein (CRP) in human serum plasma and cell culture supernatants It includes a 96-well plate with capture antibody detection antibody with an electrochemiluminescent label calibrators diluent wash buffer read buffer and plate seals The assay range is 0 016-50 ng/mL with 0 008 ng/mL sensitivity requiring only 50 L of sample It processes up to 94 samples in duplicate with high sensitivity and a wide dynamic range