Learn More
Meso Scale Discovery V-PLEX Plus Human Eotaxin Kit
SDP
Catalog No. 5000082328
Encompass
The Meso Scale Discovery V-PLEX Human Eotaxin Kit is an electrochemiluminescent immunoassay for the quantitative measurement of human Eotaxin in cell culture supernatants serum plasma and other biological fluids The kit contains a 96-well plate coated with capture antibodies detection antibodies labeled with an electrochemiluminescent compound and calibrator solutions for generating a standard curve The assay has a detection range of 0 06-2500 pg/mL and requires 25 L sample size The total assay time is 3 5 hours This highly sensitive and reproducible kit can be used to measure Eotaxin levels in studies of allergic diseases and inflammation
Catalog No. 50-000-82328 Supplier Meso Scale Discovery Supplier No. K151NSG1
The Meso Scale Discovery V-PLEX Human Eotaxin Kit is an electrochemiluminescent immunoassay for the quantitative measurement of human Eotaxin in cell culture supernatants serum plasma and other biological fluids The kit contains a 96-well plate coated with capture antibodies detection antibodies labeled with an electrochemiluminescent compound and calibrator solutions for generating a standard curve The assay has a detection range of 0 06-2500 pg/mL and requires 25 L sample size The total assay time is 3 5 hours This highly sensitive and reproducible kit can be used to measure Eotaxin levels in studies of allergic diseases and inflammation