Learn More
Meso Scale Discovery V-PLEX Plus Human Eotaxin Kit
SDP
Catalog No. 5000082329
Encompass
The V-PLEX Plus Human Eotaxin Kit from Meso Scale Discovery is a multiplex immunoassay kit that allows for the simultaneous quantitative measurement of human Eotaxin in serum plasma and cell culture supernatants The kit contains a 96-well plate coated with capture antibodies against Eotaxin detection antibodies standards buffers substrate and plate seals The electrochemiluminescent assay has high sensitivity and a broad dynamic range It requires a small sample volume and provides results in as little as 3 5 hours The kit contains sufficient reagents for one 96-well plate testing 40 samples in duplicate
Catalog No. 50-000-82329 Supplier Meso Scale Discovery Supplier No. K151NSG4
Add to Cart
Add to Cart
Request a Quote
The V-PLEX Plus Human Eotaxin Kit from Meso Scale Discovery is a multiplex immunoassay kit that allows for the simultaneous quantitative measurement of human Eotaxin in serum plasma and cell culture supernatants The kit contains a 96-well plate coated with capture antibodies against Eotaxin detection antibodies standards buffers substrate and plate seals The electrochemiluminescent assay has high sensitivity and a broad dynamic range It requires a small sample volume and provides results in as little as 3 5 hours The kit contains sufficient reagents for one 96-well plate testing 40 samples in duplicate