Learn More
Meso Scale Discovery V-PLEX Plus Human Flt-1 Kit
SDP
Catalog No. 5000079830
Encompass
The V-PLEX Plus Human FLT-1 Kit from Meso Scale Discovery is a multiplex immunoassay kit that allows for the simultaneous quantitative measurement of human FLT-1 (VEGFR-1) in serum plasma and cell culture supernatants The kit contains a 96-well plate coated with capture antibodies against FLT-1 detection antibodies conjugated to an electrochemiluminescent label calibrator solutions diluent wash buffer read buffer and plate seals The assay has a detection range of 3 5-10 000 pg/mL and exhibits minimal cross-reactivity with related analytes The kit provides all necessary reagents to perform the immunoassay on the MESO QuickPlex SQ 120 instrument
Catalog No. 50-000-79830 Supplier Meso Scale Discovery Supplier No. K151RZG4
Add to Cart
Add to Cart
Request a Quote
The V-PLEX Plus Human FLT-1 Kit from Meso Scale Discovery is a multiplex immunoassay kit that allows for the simultaneous quantitative measurement of human FLT-1 (VEGFR-1) in serum plasma and cell culture supernatants The kit contains a 96-well plate coated with capture antibodies against FLT-1 detection antibodies conjugated to an electrochemiluminescent label calibrator solutions diluent wash buffer read buffer and plate seals The assay has a detection range of 3 5-10 000 pg/mL and exhibits minimal cross-reactivity with related analytes The kit provides all necessary reagents to perform the immunoassay on the MESO QuickPlex SQ 120 instrument