Learn More
Meso Scale Discovery V-PLEX Plus Human Flt-1 Kit
SDP
Catalog No. 5000078513
Encompass
The V-PLEX Plus Human FLT-1 Kit from Meso Scale Discovery is a multiplex immunoassay that measures the human FLT-1 protein in serum plasma and cell culture supernatants The kit contains a 96-well plate coated with capture antibodies detection antibodies calibrators controls buffers and reagents needed to perform the assay The electrochemiluminescent detection method provides high sensitivity and a broad dynamic range This assay kit can be used for biomarker research related to angiogenesis cancer cardiovascular disease and inflammation The kit has a detection range of 78-10 000 pg/mL and the total assay time is 2 5 hours
Catalog No. 50-000-78513 Supplier Meso Scale Discovery Supplier No. K151RZG1
The V-PLEX Plus Human FLT-1 Kit from Meso Scale Discovery is a multiplex immunoassay that measures the human FLT-1 protein in serum plasma and cell culture supernatants The kit contains a 96-well plate coated with capture antibodies detection antibodies calibrators controls buffers and reagents needed to perform the assay The electrochemiluminescent detection method provides high sensitivity and a broad dynamic range This assay kit can be used for biomarker research related to angiogenesis cancer cardiovascular disease and inflammation The kit has a detection range of 78-10 000 pg/mL and the total assay time is 2 5 hours