Learn More
Meso Scale Discovery V-PLEX Plus Human GM-CSF Kit
SDP
Catalog No. 5000079178
Encompass
The Meso Scale Discovery MULTI-ARRAY GM-CSF Assay is an electrochemiluminescent immunoassay for the quantitative measurement of GM-CSF in serum plasma and cell culture supernatants It utilizes MSD s proprietary SULFO-TAG labels and multi-array plate technology to provide high sensitivity wide dynamic range and low sample volume requirements The assay has a lower limit of detection of 0 04 pg/mL and an upper limit of quantification of 1500 pg/mL It requires as little as 12 5 L of sample and can be read on MSD s SECTOR Imager instruments The assay is available in a 96-well plate format and includes all necessary reagents
Catalog No. 50-000-79178 Supplier Meso Scale Discovery Supplier No. K151RIG4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery MULTI-ARRAY GM-CSF Assay is an electrochemiluminescent immunoassay for the quantitative measurement of GM-CSF in serum plasma and cell culture supernatants It utilizes MSD s proprietary SULFO-TAG labels and multi-array plate technology to provide high sensitivity wide dynamic range and low sample volume requirements The assay has a lower limit of detection of 0 04 pg/mL and an upper limit of quantification of 1500 pg/mL It requires as little as 12 5 L of sample and can be read on MSD s SECTOR Imager instruments The assay is available in a 96-well plate format and includes all necessary reagents