Learn More
Meso Scale Discovery V-PLEX Plus Human GM-CSF Kit
SDP
Catalog No. 5000081294
Encompass
The Meso Scale Discovery V-PLEX Human GM-CSF Kit is an electrochemiluminescent immunoassay for the quantitative measurement of human granulocyte-macrophage colony-stimulating factor (GM-CSF) in cell culture supernates serum and plasma The assay utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and rapid detection of GM-CSF with high specificity and low background The kit includes a 96-well plate coated with capture antibody detection antibody conjugated to ruthenium calibrators and reagents required to perform the assay The lower limit of detection is 1 04 pg/mL with an upper limit of quantitation of 1500 pg/mL
Catalog No. 50-000-81294 Supplier Meso Scale Discovery Supplier No. K151RIG2
The Meso Scale Discovery V-PLEX Human GM-CSF Kit is an electrochemiluminescent immunoassay for the quantitative measurement of human granulocyte-macrophage colony-stimulating factor (GM-CSF) in cell culture supernates serum and plasma The assay utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and rapid detection of GM-CSF with high specificity and low background The kit includes a 96-well plate coated with capture antibody detection antibody conjugated to ruthenium calibrators and reagents required to perform the assay The lower limit of detection is 1 04 pg/mL with an upper limit of quantitation of 1500 pg/mL