Learn More
Meso Scale Discovery V-PLEX Plus Human GM-CSF Kit
SDP
Catalog No. 5000081681
Encompass
The V-PLEX Plus Human GM-CSF Kit from Meso Scale Discovery is a multiplex immunoassay kit for measuring human GM-CSF from 15-10 000 pg/mL with high sensitivity and reproducibility It uses electrochemiluminescence detection and provides reagents for 96 tests in a 3 5-hour protocol Suitable for serum plasma cell culture supernatants and other samples the kit includes a 96-well plate with capture antibodies detection antibodies labeled with an ECL compound and calibrator solutions
Catalog No. 50-000-81681 Supplier Meso Scale Discovery Supplier No. K151RIG1
The V-PLEX Plus Human GM-CSF Kit from Meso Scale Discovery is a multiplex immunoassay kit for measuring human GM-CSF from 15-10 000 pg/mL with high sensitivity and reproducibility It uses electrochemiluminescence detection and provides reagents for 96 tests in a 3 5-hour protocol Suitable for serum plasma cell culture supernatants and other samples the kit includes a 96-well plate with capture antibodies detection antibodies labeled with an ECL compound and calibrator solutions