Learn More
Meso Scale Discovery V-PLEX Plus Human ICAM-1 Kit
SDP
Catalog No. 5000078756
Encompass
The V-PLEX Plus Human ICAM-1 Kit is a 5-plate kit used to quantitatively measure human ICAM-1 in cell culture supernatants serum plasma and urine It utilizes electrochemiluminescence (ECL) technology and contains a V-PLEX Human ICAM-1 Plate Detection Antibody Concentrate Calibrator Blend Diluent 35 Read Buffer T (4X) and Wash Buffer This multiplex immunoassay kit has a detection range of 3 2-10 000 pg/mL and a sensitivity of 0 3 pg/mL It requires 2 5 hours to complete with only a 30 minute hands-on time
Catalog No. 50-000-78756 Supplier Meso Scale Discovery Supplier No. K151SUG2
The V-PLEX Plus Human ICAM-1 Kit is a 5-plate kit used to quantitatively measure human ICAM-1 in cell culture supernatants serum plasma and urine It utilizes electrochemiluminescence (ECL) technology and contains a V-PLEX Human ICAM-1 Plate Detection Antibody Concentrate Calibrator Blend Diluent 35 Read Buffer T (4X) and Wash Buffer This multiplex immunoassay kit has a detection range of 3 2-10 000 pg/mL and a sensitivity of 0 3 pg/mL It requires 2 5 hours to complete with only a 30 minute hands-on time