Learn More
Meso Scale Discovery V-PLEX Plus Human ICAM-1 Kit
SDP
Catalog No. 5000081708
Encompass
The Meso Scale Discovery V-PLEX Human ICAM-1 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of human ICAM-1 in serum plasma and cell culture supernatants The kit contains a 96-well plate coated with capture antibody detection antibody conjugated to an electrochemiluminescent label calibrator and buffer The assay has a detection range of 3 13-10 000 pg/mL and takes approximately 3 5 hours to complete This product allows for the sensitive and accurate quantification of ICAM-1 levels
Catalog No. 50-000-81708 Supplier Meso Scale Discovery Supplier No. K151SUG1
The Meso Scale Discovery V-PLEX Human ICAM-1 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of human ICAM-1 in serum plasma and cell culture supernatants The kit contains a 96-well plate coated with capture antibody detection antibody conjugated to an electrochemiluminescent label calibrator and buffer The assay has a detection range of 3 13-10 000 pg/mL and takes approximately 3 5 hours to complete This product allows for the sensitive and accurate quantification of ICAM-1 levels