Learn More
Meso Scale Discovery V-PLEX Plus Human IL-13 Kit
SDP
Catalog No. 5000079604
Encompass
The V-PLEX Plus Human IL-13 Kit from Meso Scale Discovery is an immunoassay that uses electrochemiluminescence (ECL) for the quantitative measurement of human interleukin 13 (IL-13) in cell culture supernatants serum plasma and other biological fluids The kit contains a 96-well plate coated with a monoclonal antibody specific for IL-13 a detection antibody conjugated to an ECL label calibrators and buffers for sample dilution The assay has a sensitivity of 0 3 pg/mL and a broad dynamic range from 0 3-2500 pg/mL It requires a low sample volume of 50 L and the total assay time is around 3 hours The ECL signal produced is proportional to the amount of IL-13 present in the sample
Catalog No. 50-000-79604 Supplier Meso Scale Discovery Supplier No. K151ODG4
Add to Cart
Add to Cart
Request a Quote
The V-PLEX Plus Human IL-13 Kit from Meso Scale Discovery is an immunoassay that uses electrochemiluminescence (ECL) for the quantitative measurement of human interleukin 13 (IL-13) in cell culture supernatants serum plasma and other biological fluids The kit contains a 96-well plate coated with a monoclonal antibody specific for IL-13 a detection antibody conjugated to an ECL label calibrators and buffers for sample dilution The assay has a sensitivity of 0 3 pg/mL and a broad dynamic range from 0 3-2500 pg/mL It requires a low sample volume of 50 L and the total assay time is around 3 hours The ECL signal produced is proportional to the amount of IL-13 present in the sample