Learn More
Meso Scale Discovery V-PLEX Plus Human IL-13 Kit
SDP
Catalog No. 5000081203
Encompass
The Meso Scale Discovery V-PLEX Plus Human IL-13 Kit is an electrochemiluminescent immunoassay for measuring human interleukin 13 (IL-13) in various biological fluids It includes a 96-well plate with a capture antibody for IL-13 a detection antibody with an electrochemiluminescent label calibrators diluent wash buffer read buffer and plate seals The assay s sensitivity is 0 3 pg/mL with a range of 0 3-2500 pg/mL using MSD s SECTOR Imager for high sensitivity and range ensuring reliable IL-13 quantification
Catalog No. 50-000-81203 Supplier Meso Scale Discovery Supplier No. K151ODG1
The Meso Scale Discovery V-PLEX Plus Human IL-13 Kit is an electrochemiluminescent immunoassay for measuring human interleukin 13 (IL-13) in various biological fluids It includes a 96-well plate with a capture antibody for IL-13 a detection antibody with an electrochemiluminescent label calibrators diluent wash buffer read buffer and plate seals The assay s sensitivity is 0 3 pg/mL with a range of 0 3-2500 pg/mL using MSD s SECTOR Imager for high sensitivity and range ensuring reliable IL-13 quantification