Learn More
Meso Scale Discovery V-PLEX Plus Human IL-13 Kit
SDP
Catalog No. 5000082269
Encompass
The Meso Scale Discovery V-PLEX Plus Human IL-13 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of human interleukin 13 (IL-13) in cell culture supernatants serum plasma and other biological fluids The kit contains a 96-well plate coated with capture antibody detection antibody conjugated to an electrochemiluminescent label calibrators diluent wash buffer read buffer and plate seals The assay has a detection range of 0 03-2500 pg/mL and takes approximately 3 5 hours to complete It utilizes MSD s proprietary SECTOR Imager for high sensitivity and a wide dynamic range The kit includes sufficient reagents for one 96-well plate
Catalog No. 50-000-82269 Supplier Meso Scale Discovery Supplier No. K151ODG2
Add to Cart
Add to Cart
The Meso Scale Discovery V-PLEX Plus Human IL-13 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of human interleukin 13 (IL-13) in cell culture supernatants serum plasma and other biological fluids The kit contains a 96-well plate coated with capture antibody detection antibody conjugated to an electrochemiluminescent label calibrators diluent wash buffer read buffer and plate seals The assay has a detection range of 0 03-2500 pg/mL and takes approximately 3 5 hours to complete It utilizes MSD s proprietary SECTOR Imager for high sensitivity and a wide dynamic range The kit includes sufficient reagents for one 96-well plate