Learn More
Meso Scale Discovery V-PLEX Plus Human IL-15 Kit
SDP
Catalog No. 5000079175
Encompass
The Meso Scale Discovery V-PLEX Plus Human IL-15 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of human interleukin 15 (IL-15) in cell culture supernatants serum plasma and other biological fluids The kit contains a 96-well plate coated with a monoclonal antibody specific for IL-15 a detection antibody conjugated to an electrochemiluminescent label calibrators diluent wash buffer read buffer and adhesive plate seals The assay has a sensitivity of 0 2 pg/mL and a broad dynamic range from 0 2-2000 pg/mL The total assay time is 3 5 hours This kit allows for the accurate and sensitive quantification of IL-15 levels
Catalog No. 50-000-79175 Supplier Meso Scale Discovery Supplier No. K151RDG4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX Plus Human IL-15 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of human interleukin 15 (IL-15) in cell culture supernatants serum plasma and other biological fluids The kit contains a 96-well plate coated with a monoclonal antibody specific for IL-15 a detection antibody conjugated to an electrochemiluminescent label calibrators diluent wash buffer read buffer and adhesive plate seals The assay has a sensitivity of 0 2 pg/mL and a broad dynamic range from 0 2-2000 pg/mL The total assay time is 3 5 hours This kit allows for the accurate and sensitive quantification of IL-15 levels