Learn More
Meso Scale Discovery V-PLEX Plus Human IL-15 Kit
SDP
Catalog No. 5000081269
Encompass
The Meso Scale Discovery V-PLEX Plus Human IL-15 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of human interleukin 15 (IL-15) in cell culture supernatants serum plasma (EDTA) and saliva The kit contains a 96-well plate coated with a monoclonal antibody to human IL-15 a vial of SULFO-TAG labeled anti-human IL-15 detection antibody assay and sample diluents calibrators and controls The assay has a sensitivity of 0 36 pg/mL and a range from 0 36-2500 pg/mL It uses MSD s proprietary SULFO-TAG technology and can be read on MSD s SECTOR Imager instruments
Catalog No. 50-000-81269 Supplier Meso Scale Discovery Supplier No. K151RDG1
The Meso Scale Discovery V-PLEX Plus Human IL-15 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of human interleukin 15 (IL-15) in cell culture supernatants serum plasma (EDTA) and saliva The kit contains a 96-well plate coated with a monoclonal antibody to human IL-15 a vial of SULFO-TAG labeled anti-human IL-15 detection antibody assay and sample diluents calibrators and controls The assay has a sensitivity of 0 36 pg/mL and a range from 0 36-2500 pg/mL It uses MSD s proprietary SULFO-TAG technology and can be read on MSD s SECTOR Imager instruments