Learn More
Meso Scale Discovery V-PLEX Plus Human IL-16 Kit
SDP
Catalog No. 5000079168
Encompass
The Meso Scale Discovery V-PLEX Plus Human IL-16 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of human interleukin 16 (IL-16) in cell culture supernatants serum plasma and other biological fluids This kit uses MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results with a detection range of 0 05-1000 pg/mL and a sample volume requirement of 25 L The assay can be fully automated and has a total assay time of approximately 3 5 hours This product can aid in the research of inflammatory and autoimmune diseases
Catalog No. 50-000-79168 Supplier Meso Scale Discovery Supplier No. K151REG2
The Meso Scale Discovery V-PLEX Plus Human IL-16 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of human interleukin 16 (IL-16) in cell culture supernatants serum plasma and other biological fluids This kit uses MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results with a detection range of 0 05-1000 pg/mL and a sample volume requirement of 25 L The assay can be fully automated and has a total assay time of approximately 3 5 hours This product can aid in the research of inflammatory and autoimmune diseases