Learn More
Meso Scale Discovery V-PLEX Plus Human IL-16 Kit
SDP
Catalog No. 5000079833
Encompass
The Meso Scale Discovery V-PLEX Plus Human IL-16 Kit is a multiplex immunoassay for quantifying human interleukin 16 (IL-16) in various biological fluids It uses MSD s SULFO-TAG labels and multi-array technology for sensitive and accurate IL-16 measurement with a high dynamic range and low sample volume The kit includes a 96-well plate with capture antibodies detection antibodies with ruthenium labels calibrators controls diluents wash buffer read buffer and plate seals The assay requires only 50 L of sample per well
Catalog No. 50-000-79833 Supplier Meso Scale Discovery Supplier No. K151REG4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX Plus Human IL-16 Kit is a multiplex immunoassay for quantifying human interleukin 16 (IL-16) in various biological fluids It uses MSD s SULFO-TAG labels and multi-array technology for sensitive and accurate IL-16 measurement with a high dynamic range and low sample volume The kit includes a 96-well plate with capture antibodies detection antibodies with ruthenium labels calibrators controls diluents wash buffer read buffer and plate seals The assay requires only 50 L of sample per well