Learn More
Meso Scale Discovery V-PLEX Plus Human IL-17A Kit
SDP
Catalog No. 5000078510
Encompass
The Meso Scale Discovery V-PLEX Plus Human IL-17A Kit is an electrochemiluminescent immunoassay for the quantitative measurement of IL-17A in human serum plasma and culture supernatants It utilizes MSD s proprietary V-PLEX technology based on electrochemiluminescence to provide high sensitivity broad dynamic range and rapid results The kit includes a plate coated with capture antibodies detection antibodies labeled with an electrochemiluminescent compound and calibrator solutions to generate a standard curve Sample incubation time is just 2 hours The kit can accurately quantify IL-17A concentrations from 0 04 pg/mL to 1000 pg/mL
Catalog No. 50-000-78510 Supplier Meso Scale Discovery Supplier No. K151RFG1
The Meso Scale Discovery V-PLEX Plus Human IL-17A Kit is an electrochemiluminescent immunoassay for the quantitative measurement of IL-17A in human serum plasma and culture supernatants It utilizes MSD s proprietary V-PLEX technology based on electrochemiluminescence to provide high sensitivity broad dynamic range and rapid results The kit includes a plate coated with capture antibodies detection antibodies labeled with an electrochemiluminescent compound and calibrator solutions to generate a standard curve Sample incubation time is just 2 hours The kit can accurately quantify IL-17A concentrations from 0 04 pg/mL to 1000 pg/mL