Learn More
Meso Scale Discovery V-PLEX Plus Human IL-17A Kit
SDP
Catalog No. 5000078566
Encompass
The Meso Scale Discovery V-PLEX Plus Human IL-17A Kit is a multiplex immunoassay that quantitatively measures human IL-17A in cell culture supernatants serum plasma and other biological fluids This 4-plex assay utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision with minimal matrix interference The kit includes a plate coated with capture antibodies detection antibodies labeled with an electrochemiluminescent compound and calibrator and control samples The assay has a detection range of 0 03-2500 pg/mL and requires 50 L or less sample volume per well
Catalog No. 50-000-78566 Supplier Meso Scale Discovery Supplier No. K151RFG4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX Plus Human IL-17A Kit is a multiplex immunoassay that quantitatively measures human IL-17A in cell culture supernatants serum plasma and other biological fluids This 4-plex assay utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision with minimal matrix interference The kit includes a plate coated with capture antibodies detection antibodies labeled with an electrochemiluminescent compound and calibrator and control samples The assay has a detection range of 0 03-2500 pg/mL and requires 50 L or less sample volume per well