Learn More
Meso Scale Discovery V-PLEX Plus Human IL-17A Kit
SDP
Catalog No. 5000081292
Encompass
The Meso Scale Discovery V-PLEX Plus Human IL-17A Kit is a highly sensitive and reproducible immunoassay for the quantitative measurement of human interleukin 17A (IL-17A) in cell culture supernatants serum plasma and other biological fluids This electrochemiluminescent assay utilizes MSD s patented SULFO-TAG labels and multi-array technology to provide superior performance and enable high throughput analysis The kit includes a 96-well plate pre-coated with capture antibody detection antibody conjugated to ruthenium label calibrators and reagents required to perform the assay The lower limit of detection is 0 04 pg/mL and the assay range is 0 08-1000 pg/mL
Catalog No. 50-000-81292 Supplier Meso Scale Discovery Supplier No. K151RFG2
Add to Cart
Add to Cart
The Meso Scale Discovery V-PLEX Plus Human IL-17A Kit is a highly sensitive and reproducible immunoassay for the quantitative measurement of human interleukin 17A (IL-17A) in cell culture supernatants serum plasma and other biological fluids This electrochemiluminescent assay utilizes MSD s patented SULFO-TAG labels and multi-array technology to provide superior performance and enable high throughput analysis The kit includes a 96-well plate pre-coated with capture antibody detection antibody conjugated to ruthenium label calibrators and reagents required to perform the assay The lower limit of detection is 0 04 pg/mL and the assay range is 0 08-1000 pg/mL