Learn More
Meso Scale Discovery V-PLEX Plus Human IL-17B Kit
SDP
Catalog No. 5000081963
Encompass
Meso Scale Diagnostics V-PLEX Plus Cytokine Panel 1 (human) Kit enables simultaneous quantitative measurement of 10 cytokines and chemokines (IFN- IL-10 IL-12p70 IL-13 IL-1 IL-2 IL-4 IL-6 IL-8 TNF- ) from a single sample Using electrochemiluminescence detection on a proprietary multi-array plate the kit includes all necessary reagents for analyzing serum plasma culture supernatants and other samples It allows efficient profiling of cytokines and chemokines involved in immune response and inflammation from a small sample volume
Catalog No. 50-000-81963 Supplier Meso Scale Discovery Supplier No. K151WOG2
Add to Cart
Add to Cart
Meso Scale Diagnostics V-PLEX Plus Cytokine Panel 1 (human) Kit enables simultaneous quantitative measurement of 10 cytokines and chemokines (IFN- IL-10 IL-12p70 IL-13 IL-1 IL-2 IL-4 IL-6 IL-8 TNF- ) from a single sample Using electrochemiluminescence detection on a proprietary multi-array plate the kit includes all necessary reagents for analyzing serum plasma culture supernatants and other samples It allows efficient profiling of cytokines and chemokines involved in immune response and inflammation from a small sample volume