Learn More
Meso Scale Discovery V-PLEX Plus Human IL-17C Kit
SDP
Catalog No. 5000081964
Encompass
The Meso Scale Discovery Human IL-17C Singleplex Assay Kit is a highly sensitive immunoassay that enables the quantitative measurement of human interleukin 17C (IL-17C) in serum plasma and cell culture supernatants This assay utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The kit includes a 96-well plate pre-coated with capture antibody detection antibody read buffer calibrator diluent wash buffer and a vial of lyophilized calibrator that must be reconstituted before use The assay can detect IL-17C in the range of 0 05-1500 pg/mL and requires only 25 L of sample per well
Catalog No. 50-000-81964 Supplier Meso Scale Discovery Supplier No. K151WPG1
The Meso Scale Discovery Human IL-17C Singleplex Assay Kit is a highly sensitive immunoassay that enables the quantitative measurement of human interleukin 17C (IL-17C) in serum plasma and cell culture supernatants This assay utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The kit includes a 96-well plate pre-coated with capture antibody detection antibody read buffer calibrator diluent wash buffer and a vial of lyophilized calibrator that must be reconstituted before use The assay can detect IL-17C in the range of 0 05-1500 pg/mL and requires only 25 L of sample per well