Learn More
Meso Scale Discovery V-PLEX Plus Human IL-17D Kit
SDP
Catalog No. 5000079516
Encompass
The Meso Scale Discovery V-PLEX Plus Human IL-17D Kit is an electrochemiluminescent immunoassay for the quantitative measurement of human interleukin 17D (IL-17D) in cell culture supernatants serum plasma and other biological fluids This kit contains a 96-well plate coated with a monoclonal antibody specific for human IL-17D a detection antibody conjugated to an electrochemiluminescent label calibrators diluents wash buffers and other reagents required to perform the assay The kit has a detection range of 0 03-2500 pg/mL and exhibits no cross-reactivity with other cytokines This assay allows for the accurate and sensitive quantification of IL-17D levels
Catalog No. 50-000-79516 Supplier Meso Scale Discovery Supplier No. K151WQG1
The Meso Scale Discovery V-PLEX Plus Human IL-17D Kit is an electrochemiluminescent immunoassay for the quantitative measurement of human interleukin 17D (IL-17D) in cell culture supernatants serum plasma and other biological fluids This kit contains a 96-well plate coated with a monoclonal antibody specific for human IL-17D a detection antibody conjugated to an electrochemiluminescent label calibrators diluents wash buffers and other reagents required to perform the assay The kit has a detection range of 0 03-2500 pg/mL and exhibits no cross-reactivity with other cytokines This assay allows for the accurate and sensitive quantification of IL-17D levels