Learn More
Meso Scale Discovery V-PLEX Plus Human IL-17D Kit
SDP
Catalog No. 5000082399
Encompass
The Meso Scale Discovery V-PLEX Plus Human Cytokine 30-Plex Kit is a multiplex immunoassay measuring 30 human cytokines and chemokines in a small sample It uses electrochemiluminescence detection for high sensitivity and broad dynamic range with low sample volume The kit includes a 96-well plate with capture antibodies labeled detection antibodies calibrators controls buffers and seals Suitable for profiling cytokines and chemokines from various samples it offers a short 3 5-hour assay time and all necessary reagents for quantitative analysis
Catalog No. 50-000-82399 Supplier Meso Scale Discovery Supplier No. K151WQG2
Add to Cart
Add to Cart
The Meso Scale Discovery V-PLEX Plus Human Cytokine 30-Plex Kit is a multiplex immunoassay measuring 30 human cytokines and chemokines in a small sample It uses electrochemiluminescence detection for high sensitivity and broad dynamic range with low sample volume The kit includes a 96-well plate with capture antibodies labeled detection antibodies calibrators controls buffers and seals Suitable for profiling cytokines and chemokines from various samples it offers a short 3 5-hour assay time and all necessary reagents for quantitative analysis