Learn More
Meso Scale Discovery V-PLEX Plus Human IL-1a Kit
SDP
Catalog No. 5000079883
Encompass
The V-PLEX Plus Human IL-1alpha Kit from Meso Scale Discovery is an immunoassay that uses electrochemiluminescence (ECL) for the quantitative measurement of human interleukin 1 alpha (IL-1alpha) in cell culture supernatants serum plasma and other biological fluids This kit includes a V-PLEX Human IL-1alpha Plate V-PLEX Detection Antibody Read Buffer and ancillary reagents required to perform testing The assay has high sensitivity and wide dynamic range with a lower limit of detection of 0 05 pg/mL and upper limit of 10 000 pg/mL It utilizes MSD s patented V-PLEX technology that provides rapid results in just 3 5 hours
Catalog No. 50-000-79883 Supplier Meso Scale Discovery Supplier No. K151RBG1
The V-PLEX Plus Human IL-1alpha Kit from Meso Scale Discovery is an immunoassay that uses electrochemiluminescence (ECL) for the quantitative measurement of human interleukin 1 alpha (IL-1alpha) in cell culture supernatants serum plasma and other biological fluids This kit includes a V-PLEX Human IL-1alpha Plate V-PLEX Detection Antibody Read Buffer and ancillary reagents required to perform testing The assay has high sensitivity and wide dynamic range with a lower limit of detection of 0 05 pg/mL and upper limit of 10 000 pg/mL It utilizes MSD s patented V-PLEX technology that provides rapid results in just 3 5 hours